Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000973.4 | NP_000964.1 | 5 | 618 | ||
NM_001317771.1 | NP_001304700.1 | 5 | 661 | |||
NM_001317782.1 | NP_001304711.1 | 4 | 786 | |||
NM_033301.2 | NP_150644.1 | 5 | 682 | |||
XM_017013723.1 | XP_016869212.1 | 5 | 593 | |||
GenBank mRNA | AK289459.1 | - | 5 | 569 | ||
BC000047.2 | - | 3 | 390 | |||
BC000077.2 | - | 5 | 553 | 3506015 | ||
BC012197.1 | - | 5 | 637 | 2905945 | ||
BC013104.1 | - | 4 | 550 | 3924874 | ||
BC030151.2 | - | 4 | 540 | |||
BC093064.1 | - | 5 | 592 | 30716964 | ||
BM783910.1 | - | 2 | 194 | |||
BQ929065.1 | - | 5 | 629 | |||
BT007379.1 | - | 4 | 526 | |||
CR457046.1 | - | 4 | 526 | |||
Z28407.1 | - | 5 | 569 |