Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011289.3 | NP_035419.1 | 2 | 284 | ||
GenBank mRNA | AF214527.1 | - | 2 | 127 | ||
AK002945.1 | - | 2 | 101 | |||
AK008120.1 | - | 2 | 109 | |||
AK010627.1 | - | 2 | 102 | |||
AK012566.1 | - | 2 | 104 | |||
AK088211.1 | - | 2 | 93 | |||
AK132027.1 | - | 2 | 112 | |||
AK146291.1 | - | 2 | 112 | |||
BC024366.1 | - | 2 | 97 | 4986323 | ||
BC082284.1 | - | 2 | 97 | 5339848 | ||
BC090395.1 | - | 2 | 88 | 5062830 | ||
BQ748836.1 | - | 2 | 89 | |||
CN831363.1 | - | 288 |