Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5830473C10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252661.1 | NP_001239590.1 | 6 | 718 | ||
GenBank mRNA | CB521232.1 | - | 3 | 299 |