Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000661.4 | NP_000652.2 | 2, 3 | 111 | ||
NM_001024921.3 | NP_001020092.1 | 2, 3 | 269 | |||
GenBank mRNA | AB062431.1 | - | 1 | 58 | ||
AK130966.1 | - | 1 | 74 | |||
AK297178.1 | - | 2, 3 | 94 | |||
AK303908.1 | - | 1 | 74 | |||
AK312241.1 | - | 1 | 78 | |||
AL039355.2 | - | 2, 3 | 244 | |||
AY320412.1 | - | 2, 3 | 94 | |||
AY376242.1 | - | 1, 2 | 228 | |||
AY597415.1 | - | 1 | 85 | |||
BC000483.2 | - | 1 | 63 | 2964733 | ||
BC004156.2 | - | 1 | 57 | 2961026 | ||
BC004206.2 | - | 1 | 57 | 2961026 | ||
BC007967.2 | - | 1 | 113 | 4304670 | ||
BC012149.1 | - | 1 | 113 | 4554838 | ||
BC031906.1 | - | 1, 2 | 58 | 4996986 | ||
BC066318.1 | - | 1 | 78 | 5277491 | ||
BC070214.1 | - | 1 | 117 | 6605055 | ||
BC100285.1 | - | 1, 2 | 218 | 6022858 | ||
BG829769.1 | - | 1, 2 | 59 | |||
BM841336.1 | - | 2, 3 | 111 | |||
D14531.1 | - | 1, 2 | 58 | |||
D16961.1 | - | 1 | 31 | |||
U09953.1 | - | 2, 3 | 81 | |||
U21138.1 | - | 1 | 58 |