Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000977.3 | NP_000968.2 | 6 | 660 | ||
NM_001243130.1 | NP_001230059.1 | 6 | 603 | |||
NM_001243131.1 | NP_001230060.1 | 7 | 519 | |||
NM_033251.2 | NP_150254.1 | 5 | 833 | |||
GenBank mRNA | AB062392.1 | - | 5 | 609 | ||
AK025405.1 | - | 1 | 654 | |||
AK026501.1 | - | 1 | 2328 | |||
AK127579.1 | - | 6 | 637 | |||
AK130963.1 | - | 6 | 556 | |||
AK130964.1 | - | 6 | 505 | |||
AK290893.1 | - | 6 | 635 | |||
AK294655.1 | - | 6 | 516 | |||
AK297198.1 | - | 7 | 495 | |||
AK308545.1 | - | 5 | 475 | |||
BC000851.1 | - | 1 | 613 | |||
BC004954.1 | - | 1 | 623 | 3623320 | ||
BC006179.1 | - | 6 | 623 | |||
BC007345.2 | - | 6 | 625 | 3688320 | ||
BC007563.1 | - | 5 | 821 | 2989021 | ||
BC007805.2 | - | 6 | 621 | 4298327 | ||
BC010994.2 | - | 5 | 616 | 3842093 | ||
BC013078.1 | - | 6 | 621 | 3459415 | ||
BC014167.2 | - | 1 | 621 | 4550361 | ||
BC020804.1 | - | 5 | 833 | 4276103 | ||
BC027463.1 | - | 6 | 621 | 5111302 | ||
BC063378.1 | - | 1 | 624 | 3542514 | ||
BC066320.1 | - | 6 | 635 | 5300601 | ||
BC093063.1 | - | 1 | 637 | 30401947 | ||
BC106058.1 | - | 6 | 660 | 4992467 | ||
BG706207.1 | - | 6 | 582 | |||
X64707.1 | - | 6 | 635 |