Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026065.3 | NP_080341.1 | 2 | 153 | ||
GenBank mRNA | AK002530.1 | - | 2 | 138 | ||
AK003427.2 | - | 2 | 154 | |||
AK012227.1 | - | 2 | 138 | |||
AK013693.1 | - | 2 | 150 | |||
AK154032.1 | - | 2 | 149 | |||
BC024337.1 | - | 2 | 134 | 4984291 |