Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110251.1 | NP_001103721.1 | 10 | 768 | ||
NM_013551.2 | NP_038579.2 | 10 | 826 | |||
GenBank mRNA | AK028234.1 | - | 10 | 694 | ||
AK143558.1 | - | 10 | 693 | |||
AK151020.1 | - | 10 | 792 | |||
AK153884.1 | - | 10 | 756 | |||
AK166734.1 | - | 10 | 724 | |||
AK167702.1 | - | 10 | 785 | |||
AK218865.1 | - | 1 | 40 | |||
BC003861.1 | - | 10 | 786 | 3487642 |