Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000984.5 | NP_000975.2 | 5 | 638 | ||
GenBank mRNA | BC058041.1 | - | 5 | 634 | 6716166 |