Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021134.3 | NP_066957.3 | 5 | 421 | ||
GenBank mRNA | AK313805.1 | - | 5 | 361 | ||
BC027710.2 | - | 5 | 336 | 4298000 | ||
U26596.1 | - | 5 | 384 | |||
Z49254.1 | - | 5 | 384 |
Gene Symbol: LOC107987373 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724896.2 | XP_006724959.1 | 5 | 421 |