Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm6497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080482.1 | - | 1201 |
Gene Symbol: Gm4635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK145383.1 | - | 3942 |
Gene Symbol: L1Md-A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U16662.1 | - | 1 | 238 | ||
U16664.1 | - | 1 | 238 | |||
U16666.1 | - | 1 | 238 | |||
U16672.1 | - | 1 | 238 |
Gene Symbol: Pnpla8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK145751.1 | - | 2002 |