Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000986.3 | NP_000977.1 | 4 | 271 | ||
GenBank mRNA | AK311992.1 | - | 4 | 275 | ||
BC000690.2 | - | 4 | 250 | 3349215 | ||
BC070193.1 | - | 4 | 248 | 6733162 | ||
BU754101.1 | - | 4 | 279 | |||
CD513036.1 | - | 3 | 257 | |||
CR456729.1 | - | 3 | 228 | |||
EU668341.1 | - | 4 | 250 | |||
M94314.1 | - | 4 | 267 |