Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026688.2 | NP_080964.1 | 6, 7 | 696 | ||
GenBank mRNA | AK002501.1 | - | 6, 7 | 669 | ||
AK088337.1 | - | 6, 7 | 697 | |||
BC027270.1 | - | 6, 7 | 650 | 3491580 | ||
BC119267.1 | - | 6, 7 | 650 | 8734016 | ||
BC119269.1 | - | 6, 7 | 651 | 8734018 |