Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037741.3 | NP_001032830.2 | 3 | 424 | ||
NM_008162.3 | NP_032188.3 | 3 | 370 | |||
GenBank mRNA | AB072797.1 | - | 3 | 406 | ||
AF045768.1 | - | 3 | 288 | |||
AF045769.1 | - | 3 | 179 | |||
AF274027.1 | - | 3 | 400 | |||
AK006441.1 | - | 3 | 265 | |||
AK160389.1 | - | 3 | 197 | |||
AK160509.1 | - | 3 | 173 | |||
AK167881.1 | - | 3 | 274 | |||
AK168027.1 | - | 3 | 197 | |||
BC083137.1 | - | 2 | 157 | |||
BC106147.1 | - | 3 | 217 | 4936866 | ||
BU961981.1 | - | 427 | ||||
BY238083.1 | - | 3 | 294 | |||
CA466677.1 | - | 3 | 351 | |||
D87896.1 | - | 3 | 370 |