Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037741.3 | NP_001032830.2 | 4 | 538 | ||
NM_008162.3 | NP_032188.3 | 4 | 484 | |||
GenBank mRNA | AB072797.1 | - | 4 | 520 | ||
AF045768.1 | - | 4 | 402 | |||
AF045769.1 | - | 4 | 293 | |||
AF274027.1 | - | 4 | 514 | |||
AK006441.1 | - | 4 | 379 | |||
AK145770.1 | - | 4 | 404 | |||
AK160389.1 | - | 4 | 311 | |||
AK160509.1 | - | 4 | 287 | |||
AK167881.1 | - | 4, 5 | 388 | |||
AK168027.1 | - | 4 | 311 | |||
BC083137.1 | - | 3 | 271 | |||
BC106147.1 | - | 4 | 331 | 4936866 | ||
BY238083.1 | - | 4 | 408 | |||
CA466677.1 | - | 4 | 465 | |||
D87896.1 | - | 4 | 484 |