Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000995.4 | NP_000986.2 | 2 | 99 | ||
NM_001319232.1 | NP_001306161.1 | 3 | 245 | |||
NM_001319234.1 | NP_001306163.1 | 2 | 229 | |||
NM_001319235.1 | NP_001306164.1 | 2 | 179 | |||
NM_001319236.1 | NP_001306165.1 | 2 | 99 | |||
NM_033625.3 | NP_296374.1 | 2 | 179 | |||
GenBank mRNA | AK055920.1 | - | 1 | 1382 | ||
BC001773.1 | - | 2 | 113 | 3355603 | ||
BC070208.1 | - | 2 | 80 | 6724916 | ||
BC106009.1 | - | 2 | 91 | 4390762 | ||
BE394007.1 | - | 2 | 145 | |||
BU588998.1 | - | 2 | 179 | |||
CB132938.1 | - | 2 | 230 | |||
CD175155.1 | - | 2 | 201 | |||
CD249923.1 | - | 2 | 137 | |||
CR542232.1 | - | 1 | 47 | |||
CR542242.1 | - | 1 | 47 | |||
DA779124.1 | - | 2 | 80 | |||
L38941.1 | - | 1 | 67 |