Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039646.2 | NP_001034735.2 | 8 | 1333 | ||
GenBank mRNA | BC112328.1 | - | 8 | 1350 | 5348193 | |
BC114209.1 | - | 8 | 1333 | 4980612 | ||
DQ985743.1 | - | 8 | 1333 |
Gene Symbol: Gbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194336.2 | NP_919317.2 | 8 | 1327 | ||
GenBank mRNA | AK129021.1 | - | 599 | |||
AK171913.1 | - | 5 | 2965 | |||
BB657433.1 | - | 2 | 391 | |||
BC007143.1 | - | 7 | 1184 | 3495728 | ||
BC052867.1 | - | 3 | 555 | |||
BC057969.1 | - | 1313 | 4978977 | |||
BC115768.1 | - | 1323 | 40085916 |