Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135657.1 | NP_001129129.1 | 24 | 3764 | ||
NM_008982.5 | NP_033008.3 | 24 | 4048 | |||
GenBank mRNA | AK131723.1 | - | 6 | 704 | ||
AK147318.1 | - | 24 | 3969 | |||
AK147556.1 | - | 24 | 3969 | |||
AK212533.1 | - | 2 | 152 | |||
AY038891.1 | - | 24 | 3866 | |||
AY039232.1 | - | 24 | 3866 | |||
D45212.1 | - | 24 | 4004 | |||
D49393.1 | - | 11 | 1021 | |||
D83203.1 | - | 24 | 3765 | |||
DQ133576.1 | - | 24 | 3779 |