Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135657.1 | NP_001129129.1 | 21 | 3362 | ||
NM_008982.5 | NP_033008.3 | 21 | 3646 | |||
GenBank mRNA | AK131723.1 | - | 3 | 302 | ||
AK147318.1 | - | 21 | 3567 | |||
AK147556.1 | - | 21 | 3567 | |||
AY038891.1 | - | 21 | 3464 | |||
AY039232.1 | - | 21 | 3464 | |||
D45212.1 | - | 21 | 3602 | |||
D49393.1 | - | 8 | 619 | |||
D83203.1 | - | 21 | 3363 | |||
DQ133576.1 | - | 21 | 3377 |