Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005.4 | NP_000996.2 | 5 | 544 | ||
NM_001256802.1 | NP_001243731.1 | 5 | 544 | |||
NM_001260506.1 | NP_001247435.1 | 5 | 592 | |||
NM_001260507.1 | NP_001247436.1 | 4 | 413 | |||
GenBank mRNA | AB062288.1 | - | 5 | 480 | ||
AK130960.1 | - | 5 | 466 | |||
AK223048.1 | - | 5 | 493 | |||
AK313051.1 | - | 5 | 513 | |||
BC003137.1 | - | 5 | 493 | 2906158 | ||
BC003577.1 | - | 5 | 485 | |||
BC013196.1 | - | 5 | 485 | |||
BC013231.1 | - | 5 | 505 | |||
BC029981.1 | - | 4 | 438 | |||
BC034149.1 | - | 5 | 517 | 4665438 | ||
BC071917.1 | - | 5 | 496 | 6297012 | ||
BC100284.1 | - | 5 | 505 | 6160514 | ||
BC150501.1 | - | 3 | 319 | |||
BE797417.1 | - | 337 | ||||
BM928330.1 | - | 5 | 574 | |||
BQ645330.1 | - | 552 | ||||
DW448503.1 | - | 1 | 34 | |||
S42658.1 | - | 5 | 495 | |||
U14990.1 | - | 5 | 487 | |||
U14991.1 | - | 5 | 487 | |||
U14992.1 | - | 5 | 487 |