Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010.2 | NP_001001.2 | 4, 5 | 532 | ||
GenBank mRNA | AB062123.1 | - | 4, 5 | 518 | ||
AK093634.1 | - | 1733 | ||||
AK291517.1 | - | 4, 5 | 531 | |||
AK311861.1 | - | 4, 5 | 529 | |||
BC000524.2 | - | 3, 4 | 494 | 2961137 | ||
BC009427.2 | - | 3, 4 | 485 | |||
BC013296.2 | - | 4, 5 | 515 | 3030319 | ||
BC027620.1 | - | 4, 5 | 535 | 4698843 | ||
BC035447.1 | - | 2, 3 | 356 | |||
BC071907.1 | - | 4, 5 | 520 | 4900361 | ||
BC071908.1 | - | 4, 5 | 520 | 5089456 | ||
BC094826.1 | - | 4, 5 | 516 | 5481378 | ||
J03537.1 | - | 3, 4 | 490 | |||
M20020.1 | - | 4, 5 | 517 |