Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010.2 | NP_001001.2 | 6 | 725 | ||
GenBank mRNA | AB062123.1 | - | 6 | 711 | ||
AK093634.1 | - | 1926 | ||||
AK291517.1 | - | 6 | 724 | |||
AK311861.1 | - | 6 | 722 | |||
BC000524.2 | - | 5 | 687 | 2961137 | ||
BC009427.2 | - | 5 | 678 | |||
BC013296.2 | - | 6 | 708 | 3030319 | ||
BC027620.1 | - | 6 | 728 | 4698843 | ||
BC035447.1 | - | 4 | 549 | |||
BC071907.1 | - | 6 | 713 | 4900361 | ||
BC071908.1 | - | 6 | 713 | 5089456 | ||
BC094826.1 | - | 6 | 709 | 5481378 | ||
J03537.1 | - | 5 | 683 | |||
M20020.1 | - | 6 | 710 |