Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008303.4 | NP_032329.1 | 4 | 717 | ||
GenBank mRNA | AK088121.1 | - | 4 | 548 | ||
AK133876.1 | - | 4 | 524 | |||
AK151935.1 | - | 4 | 527 | |||
BC024385.1 | - | 4 | 476 | 5004427 | ||
BC099385.1 | - | 4 | 528 | 30848267 | ||
CJ238411.1 | - | 3 | 401 |