Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006932.2 | NP_001006933.2 | 11 | 1284 | ||
NM_001318936.1 | NP_001305865.1 | 12 | 1335 | |||
NM_001318937.1 | NP_001305866.1 | 8 | 992 | |||
NM_001318938.1 | NP_001305867.1 | 10 | 871 | |||
NM_021135.5 | NP_066958.2 | 10 | 1137 | |||
XM_006715549.3 | XP_006715612.1 | 11 | 1166 | |||
GenBank mRNA | AB209116.1 | - | 12 | 1098 | ||
AK095751.1 | - | 11 | 1244 | |||
AK295674.1 | - | 10 | 945 | |||
AK310428.1 | - | 10 | 864 | |||
BC002363.2 | - | 12 | 1097 | 2961482 | ||
KJ535080.1 | - | 8 | 611 | |||
KJ535087.1 | - | 11 | 902 | |||
L07598.1 | - | 10 | 882 | |||
X85106.1 | - | 12 | 1053 |