Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003952.2 | NP_003943.2 | 8 | 737 | ||
XM_005274164.1 | XP_005274221.1 | 7 | 685 | |||
XM_005274165.4 | XP_005274222.1 | 7 | 562 | |||
XM_006718655.3 | XP_006718718.1 | 9 | 858 | |||
XM_006718656.3 | XP_006718719.1 | 8 | 806 | |||
XM_006718657.1 | XP_006718720.1 | 8 | 791 | |||
XM_017018108.1 | XP_016873597.1 | 9 | 869 | |||
GenBank mRNA | AB016869.1 | - | 8 | 709 | ||
AB019245.1 | - | 8 | 704 | |||
AB208827.1 | - | 4 | 4009 | |||
AF076931.1 | - | 8 | 690 | |||
AF099739.1 | - | 8 | 694 | |||
AK297527.1 | - | 9 | 843 | |||
BC000094.2 | - | 8 | 710 | 3508140 | ||
BC006106.1 | - | 8 | 693 | 2959036 | ||
BC136564.1 | - | 8 | 711 | 9020554 | ||
BX337895.2 | - | 8 | 740 |