Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mageb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028847.1 | NP_083123.1 | 2 | 576 | ||
GenBank mRNA | AK006807.1 | - | 2 | 619 | ||
BC116773.1 | - | 2 | 576 | 40126092 | ||
BC116775.1 | - | 2 | 576 | 40126094 |
Gene Symbol: Gm14781 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205268.1 | NP_001192197.1 | 2 | 621 |