Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025070.1 | NP_001020241.1 | 3 | 467 | ||
NM_001025071.1 | NP_001020242.1 | 3 | 473 | |||
NM_005617.3 | NP_005608.1 | 3 | 256 | |||
XM_017009684.1 | XP_016865173.1 | 3 | 310 | |||
GenBank mRNA | AA467888.1 | - | 1 | 23 | ||
AK312179.1 | - | 3 | 235 | |||
BC001126.1 | - | 1 | 219 | 2967098 | ||
BC003401.1 | - | 2 | 213 | 3448752 | ||
BC006784.1 | - | 1 | 224 | 2900907 | ||
BC020515.1 | - | 3 | 406 | 3856643 | ||
BC091474.1 | - | 3 | 237 | 30334486 | ||
BM454896.1 | - | 3 | 436 | |||
BM557014.1 | - | 3 | 408 |