Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025070.1 | NP_001020241.1 | 3 | 457 | ||
NM_001025071.1 | NP_001020242.1 | 3 | 463 | |||
NM_005617.3 | NP_005608.1 | 3 | 246 | |||
XM_017009684.1 | XP_016865173.1 | 3 | 300 | |||
GenBank mRNA | AK312179.1 | - | 3 | 225 | ||
BC001126.1 | - | 1 | 209 | 2967098 | ||
BC003401.1 | - | 2 | 203 | 3448752 | ||
BC006784.1 | - | 1 | 214 | 2900907 | ||
BC020515.1 | - | 3 | 396 | 3856643 | ||
BC091474.1 | - | 3 | 227 | 30334486 | ||
BM454896.1 | - | 3 | 426 | |||
BM557014.1 | - | 3 | 398 |