Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS15A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001019.4 | NP_001010.2 | 4 | 336 | ||
NM_001030009.1 | NP_001025180.1 | 4 | 270 | |||
GenBank mRNA | AB062400.1 | - | 1 | 252 | ||
AK023088.1 | - | 5 | 365 | |||
AK130967.1 | - | 4 | 270 | |||
AK291988.1 | - | 4 | 336 | |||
AK311976.1 | - | 1 | 270 | |||
AY208299.1 | - | 3 | 246 | |||
BC030569.1 | - | 3 | 268 | 5580542 | ||
BC046113.1 | - | 1 | 253 | 5736078 | ||
BC105273.1 | - | 1 | 276 | 3839617 | ||
BC105292.1 | - | 4 | 305 | 6649512 | ||
BF678907.1 | - | 4 | 337 | |||
BG035607.1 | - | 323 | ||||
X84407.1 | - | 1 | 240 |