Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025863.3 | NP_080139.3 | 3 | 1240 | ||
GenBank mRNA | AK009637.1 | - | 1 | 313 | ||
AK016642.1 | - | 3 | 1435 | |||
BC006700.1 | - | 3 | 1158 | 3501285 | ||
BC025430.1 | - | 3 | 1173 | 4017983 | ||
BC055347.1 | - | 1 | 710 |