Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lce3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270426.1 | NP_001257355.1 | 2 | 477 | ||
GenBank mRNA | EH095282.1 | - | 2 | 506 |