Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000033.3 | NP_000024.2 | 3 | 1558 | ||
GenBank mRNA | AY421736.1 | - | 3 | 689 | ||
AY421737.1 | - | 2 | 111 | |||
AY421738.1 | - | 2 | 111 | |||
BC015541.1 | - | 3 | 1539 | 3896490 | ||
BC025358.1 | - | 3 | 1558 | 4907640 | ||
Z21876.1 | - | 3 | 1545 |