Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001028.2 | NP_001019.1 | 3, 4 | 338 | ||
GenBank mRNA | AK311883.1 | - | 3, 4 | 338 | ||
BC004294.1 | - | 1 | 366 | 3622031 | ||
BC004986.1 | - | 3, 4 | 321 | 2905996 | ||
M64716.1 | - | 3, 4 | 346 |