Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172397.3 | NP_765985.1 | 5 | 397 | ||
GenBank mRNA | AK012581.1 | - | 5 | 371 | ||
AK032430.1 | - | 5 | 398 | |||
AK040486.1 | - | 5 | 398 | |||
AK049809.1 | - | 5 | 367 | |||
AK167349.1 | - | 5 | 367 | |||
AK169829.1 | - | 5 | 368 | |||
AK171794.1 | - | 5 | 676 | |||
AK188791.1 | - | 2 | 96 | |||
BC068130.1 | - | 5 | 341 | 6413206 |