Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ighv7-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X66662.1 | - | 2 | 282 |
Gene Symbol: Igh-VS107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M37671.1 | - | 1 | 152 | ||
M37673.1 | - | 1 | 152 | |||
X14096.1 | - | 2 | 209 | |||
X63806.1 | - | 1 | 152 | |||
Z22026.1 | - | 1 | 152 |
Gene Symbol: Ighv7-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HF569636.1 | - | 129 | |||
L16829.1 | - | 152 | ||||
L16831.1 | - | 152 | ||||
L30146.1 | - | 152 | ||||
U51457.1 | - | 152 | ||||
U55461.1 | - | 136 | ||||
X66655.1 | - | 2 | 280 | |||
X66657.1 | - | 2 | 280 | |||
X66658.1 | - | 2 | 280 |