Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031.4 | NP_001022.1 | 1 | 148 | ||
GenBank mRNA | AK293636.1 | - | 2 | 397 | ||
AK301638.1 | - | 2 | 397 | |||
AK311925.1 | - | 3 | 148 | |||
AU126783.1 | - | 3 | 148 | |||
BC000354.1 | - | 1 | 134 | 2964364 | ||
BC018810.1 | - | 1 | 134 | |||
BC021239.2 | - | 1 | 127 | 4309752 | ||
BC070217.1 | - | 1 | 148 | 4248660 | ||
BC070218.1 | - | 1 | 145 | 6649143 | ||
CR457055.1 | - | 1 | 117 | |||
L05091.1 | - | 3 | 132 | |||
U58682.1 | - | 3 | 129 |