Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039242.2 | NP_001034331.1 | 5 | 588 | ||
NM_172568.2 | NP_766156.2 | 5 | 588 | |||
GenBank mRNA | AK132734.1 | - | 5 | 577 | ||
BC100474.1 | - | 5 | 581 | 30918723 |