Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042576.1 | NP_001036041.1 | 11 | 1822 | ||
NM_004587.2 | NP_004578.2 | 10 | 1745 | |||
GenBank mRNA | AB037819.2 | - | 3501 | |||
AB287347.2 | - | 7 | 3157 | |||
AF006751.1 | - | 10 | 1538 | |||
AL833822.1 | - | 8 | 1533 | |||
BC000099.2 | - | 5 | 503 | |||
BC020482.1 | - | 7 | 770 | |||
BC080526.1 | - | 6 | 629 | |||
BC128577.1 | - | 10 | 1931 | 40128738 | ||
BC128578.1 | - | 9 | 965 | 40128742 |