Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028148.2 | NP_082424.2 | 11 | 2342 | ||
GenBank mRNA | AK012092.1 | - | 1 | 1187 | ||
AK129478.1 | - | 11 | 2234 | |||
BC058682.1 | - | 11 | 2323 |