Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042576.1 | NP_001036041.1 | 24 | 3026 | ||
NM_004587.2 | NP_004578.2 | 23 | 2949 | |||
GenBank mRNA | AB037819.2 | - | 4705 | |||
AB287347.2 | - | 20 | 4361 | |||
AF006751.1 | - | 23 | 2742 | |||
AK093465.1 | - | 11 | 1307 | |||
AK098319.1 | - | 4 | 1433 | |||
BC000099.2 | - | 18 | 1707 | |||
BC080526.1 | - | 19 | 1833 | |||
BC128577.1 | - | 23 | 3135 | 40128738 |