Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027191.2 | NP_081467.2 | 3 | 415 | ||
NM_028334.4 | NP_082610.1 | 3 | 433 | |||
GenBank mRNA | AK010370.1 | - | 3 | 359 | ||
AK012890.1 | - | 3 | 429 | |||
AK040463.1 | - | 3 | 416 | |||
AK165537.1 | - | 3 | 379 | |||
BC011102.1 | - | 3 | 399 | 3982076 | ||
BC032180.1 | - | 2 | 343 | 5151847 |