Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026775.4 | NP_081051.1 | 1 | 171 | ||
GenBank mRNA | AK003676.1 | - | 1 | 154 | ||
AK084117.1 | - | 1 | 154 | |||
AK150605.1 | - | 1 | 143 | |||
AK152643.1 | - | 1 | 154 | |||
AK152656.1 | - | 1 | 143 | |||
BC064755.1 | - | 1 | 145 | 30029180 | ||
BC085097.1 | - | 1 | 150 | 5349826 | ||
BY343029.1 | - | 1 | 171 |