Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021136.2 | NP_066959.1 | 8 | 2468 | ||
NM_206852.2 | NP_996734.1 | 6 | 863 | |||
XM_017021578.1 | XP_016877067.1 | 6 | 752 | |||
GenBank mRNA | AK128113.1 | - | 3 | 2328 | ||
AK290534.1 | - | 6 | 863 | |||
AK297492.1 | - | 7 | 770 | |||
BC000314.2 | - | 6 | 648 | |||
BC003003.1 | - | 6 | 648 | |||
BC090862.1 | - | 8 | 2383 | 6179803 | ||
BC111694.1 | - | 9 | 2158 | 40035553 | ||
BX449967.2 | - | 6 | 851 | |||
CR457030.1 | - | 6 | 555 | |||
L10333.1 | - | 8 | 2381 | |||
L10334.1 | - | 6 | 1289 | |||
L10335.1 | - | 6 | 583 |