Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021136.2 | NP_066959.1 | 5 | 2308 | ||
NM_206852.2 | NP_996734.1 | 3 | 703 | |||
XM_017021578.1 | XP_016877067.1 | 3 | 592 | |||
GenBank mRNA | AK128113.1 | - | 1 | 722 | ||
AK290534.1 | - | 3 | 703 | |||
AK297492.1 | - | 4 | 610 | |||
BC000314.2 | - | 3 | 488 | |||
BC003003.1 | - | 3 | 488 | |||
BC090862.1 | - | 5 | 2223 | 6179803 | ||
BC111694.1 | - | 6 | 1998 | 40035553 | ||
BX449967.2 | - | 3 | 691 | |||
CR457030.1 | - | 3 | 395 | |||
L10333.1 | - | 5 | 2221 | |||
L10334.1 | - | 3 | 1129 | |||
L10335.1 | - | 3 | 423 |