Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm14743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126321.2 | NP_001119793.1 | 2, 3 | 231 | ||
GenBank mRNA | BE647313.1 | - | 1, 2 | 33 |