Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136484.3 | NP_001129956.3 | 3 | 2067 | ||
NM_010265.5 | NP_034395.4 | 3 | 2108 | |||
NM_173442.5 | NP_775618.3 | 3 | 2140 | |||
GenBank mRNA | AK045216.1 | - | 3 | 2091 |