Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm17a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011590.2 | NP_035720.1 | 5 | 389 | ||
GenBank mRNA | AF106620.1 | - | 5 | 371 | ||
AK003114.1 | - | 5 | 396 | |||
AK003129.1 | - | 5 | 390 | |||
BC010830.1 | - | 5 | 377 | 4220707 | ||
BC098216.1 | - | 5 | 386 | 1527710 |