Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042388.2 | NP_001035847.1 | 15 | 2284 | ||
NM_005134.3 | NP_005125.1 | 15 | 2233 | |||
NR_052003.1 | 15 | 2251 | ||||
XM_011525775.2 | XP_011524077.1 | 15 | 2410 | |||
XM_011525776.1 | XP_011524078.1 | 15 | 2274 | |||
XM_011525779.2 | XP_011524081.1 | 12 | 2077 | |||
XM_017026105.1 | XP_016881594.1 | 14 | 2251 | |||
XM_017026106.1 | XP_016881595.1 | 14 | 2088 | |||
XM_017026107.1 | XP_016881596.1 | 14 | 2103 | |||
GenBank mRNA | AF100744.1 | - | 4 | 451 | ||
AF111106.1 | - | 15 | 2203 | |||
AF200478.1 | - | 15 | 2183 | |||
AK025509.1 | - | 7 | 1391 | |||
AK027064.1 | - | 15 | 2235 | |||
AK292538.1 | - | 15 | 2164 | |||
AK295533.1 | - | 6 | 1018 | |||
AK315106.1 | - | 15 | 2191 | |||
BC060829.1 | - | 15 | 2185 | 30342594 |