Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006271.1 | NP_006262.1 | 3 | 301 | ||
GenBank mRNA | AB451316.1 | - | 2 | 188 | ||
AB451446.1 | - | 2 | 188 | |||
AK126904.1 | - | 2 | 2326 | |||
AK312152.1 | - | 3 | 258 | |||
BC014392.1 | - | 3 | 263 | 3543900 | ||
BC072006.1 | - | 3 | 422 | |||
BC094843.1 | - | 3 | 424 | |||
BT006938.1 | - | 2 | 188 | |||
KU178317.1 | - | 2 | 188 | |||
KU178318.1 | - | 2 | 374 | |||
X58079.1 | - | 3 | 301 |