Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002961.2 | NP_002952.1 | 2 | 139 | ||
NM_019554.2 | NP_062427.1 | 3 | 188 | |||
GenBank mRNA | AK292083.1 | - | 2 | 139 | ||
AV735911.1 | - | 1 | 23 | |||
BC000838.1 | - | 3 | 172 | 3456833 | ||
BC016300.1 | - | 2 | 143 | 4247807 | ||
BC024974.1 | - | 2 | 123 | |||
CB109749.1 | - | 3 | 188 | |||
CR450345.1 | - | 1 | 70 | |||
M80563.1 | - | 2 | 205 |