Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002963.3 | NP_002954.2 | 2, 3 | 208 | ||
GenBank mRNA | BC034687.1 | - | 2, 3 | 176 | 4751685 | |
BF090388.1 | - | 2, 3 | 259 | |||
BX476368.1 | - | 1, 2 | 169 | |||
CR542164.1 | - | 1, 2 | 137 | |||
M86757.1 | - | 2, 3 | 186 |